Assay Details
Target Gene Details
Entrez Gene ID: | 78990 |
Gene Name: | OTU deubiquitinase, ubiquitin aldehyde binding 2 |
Gene Aliases: |
C14orf137, OTB2, OTU2 |
Location: |
Chr.14:94016083-94048933 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OTUB2 | NM_023112.3 | NP_075601.1 | ||
XM_005268055.3 | XP_005268112.1 | |||
AK025569.1 | BAB15172.1 | |||
BC009615.1 | AAH09615.1 | |||
BC068058.1 | AAH68058.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34067 | Chr.14:94024355 - 94160851 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv565620 | Chr.14:93990701 - 94031622 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)