Assay Details
Target Gene Details
Entrez Gene ID: | 11198 |
Gene Name: | SPT16 homolog, facilitates chromatin remodeling subunit |
Gene Aliases: |
CDC68, FACTP140, SPT16, SPT16/CDC68 |
Location: |
Chr.14:21351472-21384266 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SUPT16H | NM_007192.3 | NP_009123.1 | ||
XM_011536381.2 | XP_011534683.1 | |||
AK001719.1 | ||||
AK024072.1 | ||||
BC000565.2 | AAH00565.1 | |||
BC014046.2 | AAH14046.1 | |||
BC064561.1 | AAH64561.1 | |||
BC073849.1 | AAH73849.1 | |||
BC111402.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |