Assay Details
Target Gene Details
Entrez Gene ID: | 3091 |
Gene Name: | hypoxia inducible factor 1 alpha subunit |
Gene Aliases: |
HIF-1-alpha, HIF-1A, HIF-1alpha, HIF1, HIF1-ALPHA, MOP1, PASD8, bHLHe78 |
Location: |
Chr.14:61695401-61748259 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HIF1A | NM_001243084.1 | NP_001230013.1 | ||
NM_001530.3 | NP_001521.1 | |||
NM_181054.2 | NP_851397.1 | |||
AB073325.1 | BAB70608.1 | |||
AB500182.1 | ||||
AB733094.1 | ||||
AF207601.1 | AAF20139.1 | |||
AF207602.1 | AAF20140.1 | |||
AF304431.1 | AAG43026.1 | |||
AK296876.1 | ||||
AK299551.1 | ||||
AK304315.1 | ||||
AK304436.1 | ||||
AK312397.1 | ||||
AU123241.1 | ||||
BC012527.2 | AAH12527.1 | |||
BT009776.1 | AAP88778.1 | |||
BX648795.1 | ||||
DC344032.1 | ||||
FJ790247.1 | ||||
U22431.1 | AAC50152.1 | |||
U29165.1 | AAC51210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952848 | Chr.14:61696883 - 61721682 on Build GRCh38 | Duplication |
![]() |
nsv564884 | Chr.14:61667038 - 61740839 on Build GRCh38 | Loss |
![]() ![]() |
dgv3721n54 | Chr.14:61682190 - 61740839 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)