Assay Details
Target Gene Details
Entrez Gene ID: | 112849 |
Gene Name: | trans-L-3-hydroxyproline dehydratase |
Gene Aliases: |
C14orf149 |
Location: |
Chr.14:59465545-59484438 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
L3HYPDH | XM_011536386.2 | XP_011534688.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832810 | Chr.14:59317451 - 59478189 on Build GRCh38 | Loss | DAAM1 GPR135 L3HYPDH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |