Assay Details
Target Gene Details
Entrez Gene ID: | 5583 |
Gene Name: | protein kinase C eta |
Gene Aliases: |
PKC-L, PKCL, PRKCL, nPKC-eta |
Location: |
Chr.14:61280953-61550980 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRKCH | XM_017021458.1 | XP_016876947.1 | ||
AK296158.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv564881 | Chr.14:60457291 - 61377535 on Build GRCh38 | Gain | C14orf39 PRKCH LOC105378189 MNAT1 SIX4 TMEM30B SIX1 SALRNA1 SLC38A6 SIX6 TRMT5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |