Assay Details
Target Gene Details
Entrez Gene ID: | 10243 |
Gene Name: | gephyrin |
Gene Aliases: |
GEPH, GPH, GPHRYN, HKPX1, MOCODC |
Location: |
Chr.14:66507407-67735831 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPHN | XM_017020925.1 | XP_016876414.1 |
Target Gene Details
Entrez Gene ID: | 5283 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class H |
Gene Aliases: |
GPI-H |
Location: |
Chr.14:67589306-67600302 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIGH | NM_004569.3 | NP_004560.1 | ||
XM_011536838.2 | XP_011535140.1 | |||
XM_017021371.1 | XP_016876860.1 | |||
XM_017021372.1 | XP_016876861.1 | |||
AK293584.1 | ||||
AK314108.1 | ||||
BC004100.2 | AAH04100.1 | |||
BC071849.1 | AAH71849.1 | |||
BP244435.1 | ||||
BT006804.1 | AAP35450.1 | |||
L19783.1 | AAA03545.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892557 | Chr.14:67469765 - 67613847 on Build GRCh38 | Gain | GPHN PIGH PLEKHH1 TMEM229B |
nsv456331 | Chr.14:67266144 - 67637063 on Build GRCh38 | Gain | EIF2S1 MIR5694 GPHN HMGB1P34 PLEK2 PIGH MPP5 PLEKHH1 ARG2 LOC105370541 TMEM229B ATP6V1D |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |