Assay Details
Target Gene Details
Entrez Gene ID: | 79866 |
Gene Name: | bora, aurora kinase A activator |
Gene Aliases: |
C13orf34 |
Location: |
Chr.13:72727739-72756198 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BORA | NM_001286746.1 | NP_001273675.1 | ||
NM_001286747.1 | NP_001273676.1 | |||
NM_024808.3 | NP_079084.3 | |||
XM_006719868.3 | XP_006719931.2 | |||
AK291496.1 | ||||
AK298608.1 | ||||
AK300758.1 | ||||
BC025367.2 | AAH25367.2 | |||
BC044213.1 | AAH44213.1 | |||
BC056876.1 | AAH56876.2 | |||
BM923278.1 | ||||
BX648605.1 | ||||
EU834129.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1049859 | Chr.13:72741128 - 72872273 on Build GRCh38 | Loss | DIS3 BORA PIBF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |