Assay Details
Target Gene Details
Entrez Gene ID: | 8600 |
Gene Name: | tumor necrosis factor superfamily member 11 |
Gene Aliases: |
CD254, ODF, OPGL, OPTB2, RANKL, TNLG6B, TRANCE, hRANKL2, sOdf |
Location: |
Chr.13:42562736-42608013 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNFSF11 | NM_003701.3 | NP_003692.1 | ||
NM_033012.3 | NP_143026.1 | |||
XM_011535280.2 | XP_011533582.1 | |||
XM_017020802.1 | XP_016876291.1 | |||
XM_017020803.1 | XP_016876292.1 | |||
AB037599.1 | BAA90488.1 | |||
AB061227.1 | BAB71768.1 | |||
AB064268.1 | BAB79693.1 | |||
AB064269.1 | BAB79694.1 | |||
AB064270.1 | BAB79695.1 | |||
AF013171.1 | AAC51762.1 | |||
AF019047.1 | AAB86811.1 | |||
AF053712.1 | AAC39731.1 | |||
BC074823.2 | AAH74823.1 | |||
BC074890.2 | AAH74890.1 | |||
BC117286.1 | ||||
BC117288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832593 | Chr.13:42571804 - 42735651 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)