Assay Details
Target Gene Details
Entrez Gene ID: | 57696 |
Gene Name: | DEAD-box helicase 55 |
Gene Aliases: |
- |
Location: |
Chr.12:123602077-123620943 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX55 | NM_020936.2 | NP_065987.1 | ||
NR_135104.1 | ||||
NR_135105.1 | ||||
XM_011538608.2 | XP_011536910.1 | |||
XM_017019710.1 | XP_016875199.1 | |||
XM_017019711.1 | XP_016875200.1 | |||
XM_017019712.1 | XP_016875201.1 | |||
XM_017019713.1 | XP_016875202.1 | |||
XM_017019714.1 | XP_016875203.1 | |||
XM_017019715.1 | XP_016875204.1 | |||
XM_017019716.1 | XP_016875205.1 | |||
XM_017019717.1 | XP_016875206.1 | |||
XM_017019718.1 | XP_016875207.1 | |||
XM_017019719.1 | XP_016875208.1 | |||
XM_017019720.1 | XP_016875209.1 | |||
XM_017019721.1 | XP_016875210.1 | |||
AB046815.1 | BAB13421.1 | |||
AK074219.1 | BAB85021.1 | |||
AK131442.1 | ||||
AK301043.1 | ||||
AK307384.1 | ||||
AL833756.1 | CAH56233.1 | |||
BC030020.2 | AAH30020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1566n100 | Chr.12:123610211 - 123711309 on Build GRCh38 | Gain | EIF2B1 TCTN2 GTF2H3 DDX55 |
nsv1042181 | Chr.12:123552853 - 123668403 on Build GRCh38 | Gain | EIF2B1 LOC101927415 TMED2 GTF2H3 DDX55 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |