Assay Details
Target Gene Details
Entrez Gene ID: | 253827 |
Gene Name: | methionine sulfoxide reductase B3 |
Gene Aliases: |
DFNB74 |
Location: |
Chr.12:65278643-65466907 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MSRB3 | NM_001031679.2 | NP_001026849.1 | ||
NM_001193460.1 | NP_001180389.1 | |||
NM_001193461.1 | NP_001180390.1 | |||
NM_198080.3 | NP_932346.1 | |||
AK293084.1 | ||||
AK299065.1 | ||||
AK316365.1 | ||||
AL833622.1 | ||||
AY358229.1 | AAQ88596.1 | |||
BC040053.1 | AAH40053.1 | |||
BX500552.1 | ||||
BX648776.1 | CAI46018.1 | |||
DC357690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832445 | Chr.12:65107107 - 65293731 on Build GRCh38 | Gain | MSRB3 WIF1 LEMD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |