Assay Details
Target Gene Details
Entrez Gene ID: | 10728 |
Gene Name: | prostaglandin E synthase 3 |
Gene Aliases: |
P23, TEBP, cPGES |
Location: |
Chr.12:56663341-56689250 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTGES3 | NM_001282601.1 | NP_001269530.1 | ||
NM_001282602.1 | NP_001269531.1 | |||
NM_001282603.1 | NP_001269532.1 | |||
NM_001282604.1 | NP_001269533.1 | |||
NM_001282605.1 | NP_001269534.1 | |||
NM_006601.6 | NP_006592.3 | |||
NR_104219.1 | ||||
XM_005268576.4 | XP_005268633.1 | |||
XM_006719199.2 | XP_006719262.1 | |||
XM_011537773.2 | XP_011536075.1 | |||
XM_011537774.2 | XP_011536076.1 | |||
XM_017018716.1 | XP_016874205.1 | |||
AK098214.1 | ||||
AK291945.1 | ||||
AK293139.1 | ||||
AK295208.1 | ||||
AK298147.1 | ||||
AK298160.1 | ||||
AL603017.1 | ||||
BC003005.1 | AAH03005.1 | |||
BM541503.1 | ||||
BP194632.1 | ||||
BU167210.1 | ||||
L24804.1 | AAA18537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | ARHGAP9 MIP SPRYD4 BAZ2A DDIT3 ANKRD52 LRP1 GLI1 NACA MARS STAT2 PIP4K2C ATP5B SLC26A10 NEMP1 APOF COQ10A ARHGEF25 PAN2 LRP1-AS STAC3 GPR182 PRIM1 R3HDM2 MIR1228 GLS2 NABP2 SHMT2 B4GALNT1 SNORD59B DTX3 OS9 NDUFA4L2 RBMS2 SDR9C7 LOC101927583 MIR6758 STAT6 DCTN2 MIR616 ZBTB39 TAC3 TIMELESS MYO1A RDH16 NXPH4 NAB2 RNF41 CNPY2 SNORD59A SNORA105C HBCBP KIF5A PTGES3 CS SLC39A5 INHBC IL23A HSD17B6 INHBE MBD6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |