Assay Details
Target Gene Details
Entrez Gene ID: | 56341 |
Gene Name: | protein arginine methyltransferase 8 |
Gene Aliases: |
HRMT1L3, HRMT1L4 |
Location: |
Chr.12:3381349-3593973 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRMT8 | NM_001256536.1 | NP_001243465.1 | ||
AK315619.1 | ||||
KR014345.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss | TIGAR CCND2-AS1 THCAT155 TSPAN9 KCNA1 PRMT8 LOC100128253 LOC101929549 KCNA5 C12orf4 LOC105369606 DYRK4 FGF23 LOC101929584 CCND2 FGF6 GALNT8 RAD51AP1 KCNA6 CRACR2A AKAP3 PARP11 NDUFA9 |
nsv580 | Chr.12:3430854 - 3471028 on Build GRCh38 | Insertion | PRMT8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |