Assay Details
Target Gene Details
Entrez Gene ID: | 10728 |
Gene Name: | prostaglandin E synthase 3 |
Gene Aliases: |
P23, TEBP, cPGES |
Location: |
Chr.12:56663341-56689250 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTGES3 | NM_001282601.1 | NP_001269530.1 | ||
NM_001282602.1 | NP_001269531.1 | |||
NM_001282603.1 | NP_001269532.1 | |||
NM_001282604.1 | NP_001269533.1 | |||
NM_001282605.1 | NP_001269534.1 | |||
NM_006601.6 | NP_006592.3 | |||
NR_104219.1 | ||||
XM_005268576.4 | XP_005268633.1 | |||
XM_006719199.2 | XP_006719262.1 | |||
XM_011537773.2 | XP_011536075.1 | |||
XM_011537774.2 | XP_011536076.1 | |||
XM_017018716.1 | XP_016874205.1 | |||
AK098214.1 | ||||
AK291945.1 | ||||
AK293139.1 | ||||
AK295208.1 | ||||
AK298147.1 | ||||
AK298160.1 | ||||
AL603017.1 | ||||
BC003005.1 | AAH03005.1 | |||
BM541503.1 | ||||
BP194632.1 | ||||
BU167210.1 | ||||
L24804.1 | AAA18537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | IL23A APOF STAC3 ARHGEF25 DCTN2 MIP OS9 INHBC RNF41 NACA SLC26A10 PAN2 MIR616 COQ10A MIR6758 LRP1 MIR1228 ATP5B NABP2 GLI1 LRP1-AS SNORD59B B4GALNT1 BAZ2A STAT6 SHMT2 LOC101927583 GPR182 HSD17B6 NEMP1 MYO1A SNORD59A SLC39A5 NAB2 MARS KIF5A ANKRD52 ARHGAP9 NXPH4 SNORA105C ZBTB39 SDR9C7 TIMELESS SPRYD4 GLS2 DDIT3 PTGES3 INHBE DTX3 RDH16 RBMS2 R3HDM2 TAC3 CNPY2 NDUFA4L2 HBCBP MBD6 CS STAT2 PRIM1 PIP4K2C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |