Assay Details
Target Gene Details
Entrez Gene ID: | 3479 |
Gene Name: | insulin like growth factor 1 |
Gene Aliases: |
IGF-I, IGFI, MGF |
Location: |
Chr.12:102395867-102481839 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGF1 | NM_000618.4 | NP_000609.1 | ||
NM_001111283.2 | NP_001104753.1 | |||
NM_001111284.1 | NP_001104754.1 | |||
XM_017019262.1 | XP_016874751.1 | |||
XM_017019263.1 | XP_016874752.1 | |||
AB209184.1 | BAD92421.1 | |||
AK312231.1 | ||||
BC152321.1 | ||||
CR541861.1 | CAG46659.1 | |||
M27544.1 | AAA52787.1 | |||
M29644.1 | AAA52543.1 | |||
M37484.1 | AAA52789.1 | |||
U40870.1 | AAA96152.1 | |||
X00173.1 | CAA24998.1 | |||
X56773.1 | CAA40092.1 | |||
X57025.1 | CAA40342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832498 | Chr.12:102334798 - 102519791 on Build GRCh38 | Gain | IGF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |