Assay Details
Target Gene Details
Entrez Gene ID: | 283383 |
Gene Name: | adhesion G protein-coupled receptor D1 |
Gene Aliases: |
GPR133, PGR25 |
Location: |
Chr.12:130953907-131141469 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRD1 | NM_198827.3 | NP_942122.2 | ||
XM_005253566.2 | XP_005253623.1 | |||
XM_011538203.2 | XP_011536505.1 | |||
XM_011538204.2 | XP_011536506.1 | |||
XM_011538205.2 | XP_011536507.1 | |||
XM_011538206.2 | XP_011536508.1 | |||
XM_011538207.2 | XP_011536509.1 | |||
XM_011538208.2 | XP_011536510.1 | |||
XM_011538209.2 | XP_011536511.1 | |||
XM_011538210.2 | XP_011536512.1 | |||
AY532280.1 | AAS37682.1 | |||
BC112307.1 | AAI12308.1 | |||
BC112309.1 | AAI12310.1 | |||
BC143775.1 | ||||
BX649116.1 | ||||
EU523118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1038008 | Chr.12:130928831 - 131167119 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv560769 | Chr.12:130916107 - 131621821 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)