Assay Details
Target Gene Details
Entrez Gene ID: | 1240 |
Gene Name: | chemerin chemokine-like receptor 1 |
Gene Aliases: |
CHEMERINR, ChemR23, DEZ, RVER1 |
Location: |
Chr.12:108288044-108339348 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMKLR1 | NM_001142343.1 | NP_001135815.1 | ||
NM_001142344.1 | NP_001135816.1 | |||
NM_001142345.1 | NP_001135817.1 | |||
NM_004072.2 | NP_004063.1 | |||
XM_017018820.1 | XP_016874309.1 | |||
AK291800.1 | ||||
BC106927.2 | AAI06928.1 | |||
BC106928.1 | AAI06929.1 | |||
BP328503.1 | ||||
DA466082.1 | ||||
DA939526.1 | ||||
U79526.1 | AAC51258.1 | |||
U79527.1 | AAC51259.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |