Assay Details
Target Gene Details
Entrez Gene ID: | 6660 |
Gene Name: | SRY-box 5 |
Gene Aliases: |
L-SOX5, L-SOX5B, L-SOX5F, LAMSHF |
Location: |
Chr.12:23529499-24562701 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOX5 | XM_011520831.2 | 15 | 4452 | XP_011519133.1 |
XM_011520832.2 | 15 | 4564 | XP_011519134.1 | |
XM_011520833.2 | 15 | 4893 | XP_011519135.1 | |
XM_011520834.2 | 19 | 5026 | XP_011519136.1 | |
XM_011520835.2 | 18 | 5078 | XP_011519137.1 | |
XM_011520837.2 | 15 | 4635 | XP_011519139.1 | |
XM_011520838.2 | 15 | 4455 | XP_011519140.1 | |
XM_011520842.2 | 8 | 3515 | XP_011519144.1 | |
XM_017019888.1 | 16 | 4907 | XP_016875377.1 | |
XM_017019889.1 | 16 | 4903 | XP_016875378.1 | |
XM_017019890.1 | 19 | 5042 | XP_016875379.1 | |
XM_017019891.1 | 19 | 4900 | XP_016875380.1 | |
XM_017019892.1 | 20 | 5121 | XP_016875381.1 | |
XM_017019893.1 | 17 | 4981 | XP_016875382.1 | |
XM_017019894.1 | 15 | 6911 | XP_016875383.1 | |
XM_017019895.1 | 15 | 4478 | XP_016875384.1 | |
XM_017019896.1 | 19 | 5039 | XP_016875385.1 | |
XM_017019897.1 | 19 | 4937 | XP_016875386.1 | |
XM_017019898.1 | 19 | 4934 | XP_016875387.1 | |
XM_017019899.1 | 18 | 4646 | XP_016875388.1 | |
XM_017019900.1 | 15 | 5260 | XP_016875389.1 | |
XM_017019901.1 | 15 | 9716 | XP_016875390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv91n21 | Chr.12:22914368 - 23536367 on Build GRCh38 | Gain | SOX5 LOC101928441 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |