Assay Details
Target Gene Details
Entrez Gene ID: | 5939 |
Gene Name: | RNA binding motif single stranded interacting protein 2 |
Gene Aliases: |
SCR3 |
Location: |
Chr.12:56521812-56596196 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBMS2 | NM_002898.3 | NP_002889.1 | ||
XM_005269059.4 | XP_005269116.1 | |||
XM_005269060.4 | XP_005269117.1 | |||
XM_005269061.3 | XP_005269118.1 | |||
XM_005269066.4 | XP_005269123.1 | |||
XM_006719541.3 | XP_006719604.1 | |||
XM_006719542.3 | XP_006719605.1 | |||
XM_006719543.3 | XP_006719606.1 | |||
XM_006719544.3 | XP_006719607.1 | |||
XM_011538638.2 | XP_011536940.1 | |||
XM_011538639.2 | XP_011536941.1 | |||
XM_011538640.2 | XP_011536942.1 | |||
AK293315.1 | ||||
AK299580.1 | ||||
AK304540.1 | ||||
AK308885.1 | ||||
BC027863.1 | AAH27863.1 | |||
BC072679.1 | AAH72679.1 | |||
D28483.1 | BAA05842.1 | |||
DC382164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | RNF41 SNORD59A LOC101927583 INHBE PIP4K2C IL23A STAT6 BAZ2A PAN2 TAC3 GPR182 NDUFA4L2 SPRYD4 PRIM1 MIP KIF5A DCTN2 LRP1 SLC26A10 NAB2 SNORD59B STAC3 ZBTB39 MARS ARHGAP9 ARHGEF25 RBMS2 CNPY2 MYO1A DDIT3 COQ10A DTX3 NACA OS9 NEMP1 MIR616 MIR6758 HSD17B6 GLI1 MBD6 SDR9C7 APOF SLC39A5 B4GALNT1 R3HDM2 ANKRD52 RDH16 STAT2 NXPH4 CS SNORA105C LRP1-AS NABP2 GLS2 TIMELESS INHBC MIR1228 PTGES3 SHMT2 HBCBP ATP5B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |