Assay Details
Target Gene Details
Entrez Gene ID: | 55884 |
Gene Name: | WD repeat and SOCS box containing 2 |
Gene Aliases: |
SBA2 |
Location: |
Chr.12:118032687-118062430 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WSB2 | NM_001278557.1 | NP_001265486.1 | ||
NM_001278558.1 | NP_001265487.1 | |||
NM_018639.4 | NP_061109.1 | |||
XM_017019642.1 | XP_016875131.1 | |||
AF163324.1 | AAF80478.1 | |||
AF229181.1 | AAF71302.1 | |||
AK294230.1 | ||||
AK295555.1 | ||||
AK298513.1 | ||||
AK310892.1 | ||||
BC015887.2 | AAH15887.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3581916 | Chr.12:117356518 - 118228514 on Build GRCh38 | Gain | VSIG10 WSB2 KSR2 RFC5 PEBP1 TAOK3 NOS1 |
nsv560388 | Chr.12:117953795 - 118044547 on Build GRCh38 | Loss | WSB2 KSR2 RFC5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |