Assay Details
Target Gene Details
Entrez Gene ID: | 84190 |
Gene Name: | methyltransferase like 25 |
Gene Aliases: |
C12orf26 |
Location: |
Chr.12:82358497-82479239 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
METTL25 | NM_001319675.1 | NP_001306604.1 | ||
NM_032230.2 | NP_115606.2 | |||
XM_005269186.4 | XP_005269243.2 | |||
XM_011538827.2 | XP_011537129.1 | |||
XM_011538828.2 | XP_011537130.1 | |||
XM_011538829.2 | XP_011537131.1 | |||
XM_017020020.1 | XP_016875509.1 | |||
XM_017020021.1 | XP_016875510.1 | |||
AK026442.1 | BAB15484.1 | |||
AK310231.1 | ||||
BC029120.1 | AAH29120.1 | |||
DB059865.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv559539 | Chr.12:81887018 - 82502439 on Build GRCh38 | Gain | LOC101928449 METTL25 CCDC59 |
nsv559541 | Chr.12:82321431 - 82714616 on Build GRCh38 | Gain | METTL25 CCDC59 TMTC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |