Assay Details
Target Gene Details
Entrez Gene ID: | 84530 |
Gene Name: | serine/arginine repetitive matrix 4 |
Gene Aliases: |
KIAA1853, MU-MB-2.76, nSR100 |
Location: |
Chr.12:118981495-119163051 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SRRM4 | NM_194286.3 | NP_919262.2 | ||
AB058756.1 | ||||
AK291361.1 | ||||
BC152471.1 | ||||
BC167162.1 | ||||
BC167163.1 | ||||
KC995185.1 | ||||
KC995186.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |