Assay Details
Target Gene Details
Entrez Gene ID: | 56341 |
Gene Name: | protein arginine methyltransferase 8 |
Gene Aliases: |
HRMT1L3, HRMT1L4 |
Location: |
Chr.12:3381349-3593973 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRMT8 | NM_001256536.1 | NP_001243465.1 | ||
AK315619.1 | ||||
KR014345.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss | PARP11 CCND2 KCNA6 PRMT8 KCNA5 LOC101929584 AKAP3 C12orf4 KCNA1 DYRK4 NDUFA9 RAD51AP1 LOC100128253 TSPAN9 THCAT155 CCND2-AS1 GALNT8 FGF23 FGF6 LOC105369606 TIGAR LOC101929549 CRACR2A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |