Assay Details
Target Gene Details
Entrez Gene ID: | 283420 |
Gene Name: | C-type lectin domain family 9 member A |
Gene Aliases: |
CD370, DNGR-1, DNGR1, UNQ9341 |
Location: |
Chr.12:10030677-10066030 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLEC9A | NM_207345.3 | NP_997228.1 | ||
AY358265.1 | AAQ88632.1 | |||
EU339276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524870 | Chr.12:10057112 - 10067286 on Build GRCh38 | Gain | CLEC9A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |