Assay Details
Target Gene Details
Entrez Gene ID: | 4973 |
Gene Name: | oxidized low density lipoprotein receptor 1 |
Gene Aliases: |
CLEC8A, LOX1, LOXIN, SCARE1, SLOX1 |
Location: |
Chr.12:10158300-10172191 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OLR1 | NM_001172632.1 | NP_001166103.1 | ||
NM_001172633.1 | NP_001166104.1 | |||
NM_002543.3 | NP_002534.1 | |||
AB010710.1 | BAA24580.1 | |||
AB102861.1 | BAC81565.1 | |||
AF035776.1 | AAC82329.1 | |||
AK292124.1 | ||||
AK295409.1 | ||||
AK298040.1 | ||||
AK308009.1 | ||||
AL551100.3 | ||||
AX429248.1 | CAD36113.1 | |||
BC022295.1 | AAH22295.1 | |||
BP219342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3628528 | Chr.12:10157803 - 10176482 on Build GRCh38 | Loss | OLR1 TMEM52B |
esv3628526 | Chr.12:10143326 - 10179529 on Build GRCh38 | Loss | OLR1 TMEM52B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |