Assay Details
Target Gene Details
Entrez Gene ID: | 283420 |
Gene Name: | C-type lectin domain family 9 member A |
Gene Aliases: |
CD370, DNGR-1, DNGR1, UNQ9341 |
Location: |
Chr.12:10030677-10066030 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLEC9A | NM_207345.3 | NP_997228.1 | ||
AK307335.1 | ||||
AY358265.1 | AAQ88632.1 | |||
DB090462.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |