Assay Details
Target Gene Details
Entrez Gene ID: | 4908 |
Gene Name: | neurotrophin 3 |
Gene Aliases: |
HDNF, NGF-2, NGF2, NT-3, NT3 |
Location: |
Chr.12:5432114-5495299 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NTF3 | NM_001102654.1 | NP_001096124.1 | ||
XM_011520963.2 | XP_011519265.1 | |||
AK293895.1 | ||||
BU939830.1 | ||||
CN267386.1 | ||||
X53655.1 | CAA37703.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468988 | Chr.12:5413390 - 6141521 on Build GRCh38 | Gain | VWF NTF3 ANO2 |
nsv1040098 | Chr.12:3987637 - 5646161 on Build GRCh38 | Gain | LOC101929549 KCNA1 FGF23 KCNA6 KCNA5 CCND2 RAD51AP1 C12orf4 NDUFA9 NTF3 DYRK4 LOC101929584 ANO2 AKAP3 FGF6 TIGAR CCND2-AS1 GALNT8 |
nsv523152 | Chr.12:5428946 - 5588893 on Build GRCh38 | Gain | NTF3 ANO2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |