Assay Details
Target Gene Details
Entrez Gene ID: | 57494 |
Gene Name: | ribosomal modification protein rimK like family member B |
Gene Aliases: |
FAM80B, NAAGS, NAAGS-I |
Location: |
Chr.12:8681606-8783098 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RIMKLB | NR_123740.1 | |||
XM_017019684.1 | XP_016875173.1 | |||
XM_017019689.1 | XP_016875178.1 | |||
XM_017019697.1 | XP_016875186.1 | |||
DB265022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain | CLEC4E KLRF1 RIMKLB A2M CLEC2B DDX12P OVOS CD69 A2MP1 AICDA LINC00987 LOC642846 LOC105376678 KLRG1 LOC101930452 LINC00612 LOC728715 M6PR KLRB1 PZP A2ML1 CLEC6A MIR1244-3 CLEC2A CLEC12A CLEC2D LOC100506159 LOC374443 A2M-AS1 LOC105369649 PHC1 LOC105369728 LOC101928030 LOC400002 KLRF2 MFAP5 CLEC4D CLECL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |