Assay Details
Target Gene Details
Entrez Gene ID: | 120939 |
Gene Name: | transmembrane protein 52B |
Gene Aliases: |
C12orf59 |
Location: |
Chr.12:10170599-10191804 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM52B | NM_001079815.1 | NP_001073283.1 | ||
DA156941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3628528 | Chr.12:10157803 - 10176482 on Build GRCh38 | Loss | TMEM52B OLR1 |
esv3628526 | Chr.12:10143326 - 10179529 on Build GRCh38 | Loss | TMEM52B OLR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |