Assay Details
Target Gene Details
Entrez Gene ID: | 144608 |
Gene Name: | chromosome 12 open reading frame 60 |
Gene Aliases: |
- |
Location: |
Chr.12:14567991-14823858 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C12orf60 |
Target Gene Details
Entrez Gene ID: | 2984 |
Gene Name: | guanylate cyclase 2C |
Gene Aliases: |
DIAR6, GUC2C, MECIL, MUCIL, STAR |
Location: |
Chr.12:14612632-14696625 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GUCY2C | NM_004963.3 | NP_004954.2 | ||
XM_011520631.2 | XP_011518933.1 | |||
BC136544.1 | ||||
BC136545.1 | ||||
M73489.1 | AAA36655.1 | |||
S57551.1 | AAB19934.2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |