Assay Details
Target Gene Details
Entrez Gene ID: | 23446 |
Gene Name: | solute carrier family 44 member 1 |
Gene Aliases: |
CD92, CDW92, CHTL1, CTL1 |
Location: |
Chr.9:105244608-105438504 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC44A1 | NM_001286730.1 | NP_001273659.1 | ||
NM_080546.4 | NP_536856.2 | |||
XM_005251855.4 | XP_005251912.1 | |||
XM_017014559.1 | XP_016870048.1 | |||
XM_017014560.1 | XP_016870049.1 | |||
AJ272365.1 | CAC82175.1 | |||
AJ420812.1 | CAD12764.1 | |||
BC049203.1 | AAH49203.1 | |||
CD101684.1 | ||||
CF994263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831678 | Chr.9:105161241 - 105350532 on Build GRCh38 | Gain+Loss | SLC44A1 |
esv3891706 | Chr.9:104480016 - 105304573 on Build GRCh38 | Gain | SLC44A1 LOC105376196 OR13C2 OR13C8 OR13C3 NIPSNAP3A OR13C5 NIPSNAP3B OR13D1 OR13C9 OR13C4 OR13F1 ABCA1 |
esv3891705 | Chr.9:104391828 - 106660921 on Build GRCh38 | Loss | SLC44A1 LOC105376196 OR13C2 FKTN OR13C8 RALGAPA1P1 TAL2 OR13C3 NIPSNAP3A OR13C5 FSD1L NIPSNAP3B LINC01505 OR13D1 OR13C9 OR13C4 OR13F1 ABCA1 TMEM38B MIR8081 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |