Assay Details
Target Gene Details
Entrez Gene ID: | 79048 |
Gene Name: | SECIS binding protein 2 |
Gene Aliases: |
SBP2 |
Location: |
Chr.9:89318473-89367117 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3621014 | Chr.9:88877891 - 89641453 on Build GRCh38 | Gain | SECISBP2 SHC3 CKS2 C9orf47 SEMA4D MIR3153 S1PR3 GADD45G UNQ6494 |
nsv1041327 | Chr.9:89298100 - 89438209 on Build GRCh38 | Gain | SECISBP2 CKS2 SEMA4D MIR3153 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |