Assay Details
Target Gene Details
Entrez Gene ID: | 4702 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A8 |
Gene Aliases: |
CI-19KD, CI-PGIV, PGIV |
Location: |
Chr.9:122132466-122159819 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA8 | NM_001318195.1 | NP_001305124.1 | ||
AK301520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831715 | Chr.9:122066612 - 122226720 on Build GRCh38 | Gain | TTLL11 MORN5 LHX6 MIR4478 NDUFA8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |