Assay Details
Target Gene Details
Entrez Gene ID: | 57720 |
Gene Name: | G protein-coupled receptor 107 |
Gene Aliases: |
GCDRP, LUSTR1, bA138E2.2 |
Location: |
Chr.9:130053706-130140169 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR107 | NM_001136557.1 | NP_001130029.1 | ||
NM_001136558.1 | NP_001130030.1 | |||
NM_001287346.1 | NP_001274275.1 | |||
NM_020960.4 | NP_066011.2 | |||
XM_017014953.1 | XP_016870442.1 | |||
XM_017014954.1 | XP_016870443.1 | |||
AB046844.1 | BAB13450.1 | |||
AF376725.1 | AAK57695.1 | |||
AK026244.1 | BAB15408.1 | |||
AK225456.1 | ||||
AK293092.1 | ||||
BC110518.2 | AAI10519.1 | |||
BC143655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1046307 | Chr.9:130029993 - 130219186 on Build GRCh38 | Gain | LOC401554 GPR107 NCS1 FNBP1 |
nsv831732 | Chr.9:129920648 - 130103900 on Build GRCh38 | Loss | GPR107 FNBP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |