Assay Details
Target Gene Details
Entrez Gene ID: | 79886 |
Gene Name: | caspase activity and apoptosis inhibitor 1 |
Gene Aliases: |
C9orf82, CAAP |
Location: |
Chr.9:26840685-26892828 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CAAP1 | NM_001167575.1 | NP_001161047.1 | ||
NM_024828.3 | NP_079104.3 | |||
AK023719.1 | BAB14655.1 | |||
AK300739.1 | ||||
AK301673.1 | ||||
BC014658.1 | AAH14658.1 | |||
BC071953.1 | AAH71953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1026827 | Chr.9:26693517 - 26904701 on Build GRCh38 | Gain | CAAP1 PLAA |
nsv1015259 | Chr.9:26347305 - 27643043 on Build GRCh38 | Gain | IFT74 CAAP1 TEK LRRC19 IFNK MOB3B IFT74-AS1 C9orf72 EQTN PLAA LINC00032 |
esv3891647 | Chr.9:26743024 - 27123262 on Build GRCh38 | Gain | IFT74 CAAP1 TEK LRRC19 IFT74-AS1 PLAA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |