Assay Details
Target Gene Details
Entrez Gene ID: | 56995 |
Gene Name: | tubby like protein 4 |
Gene Aliases: |
TUSP |
Location: |
Chr.6:158232192-158511828 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TULP4 | XM_011535946.1 | XP_011534248.1 | ||
XM_017011063.1 | XP_016866552.1 | |||
XM_017011064.1 | XP_016866553.1 | |||
XM_017011068.1 | XP_016866557.1 | |||
XM_017011069.1 | XP_016866558.1 | |||
XM_017011070.1 | XP_016866559.1 | |||
XM_017011071.1 | XP_016866560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830849 | Chr.6:158118191 - 158279850 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv3890972 | Chr.6:158082895 - 158303958 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv823889 | Chr.6:158193506 - 158261764 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)