Assay Details
Target Gene Details
Entrez Gene ID: | 654 |
Gene Name: | bone morphogenetic protein 6 |
Gene Aliases: |
VGR, VGR1 |
Location: |
Chr.6:7726778-7881728 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BMP6 | NM_001718.4 | NP_001709.1 | ||
AK300628.1 | ||||
BC096084.2 | AAH96084.1 | |||
BC096085.2 | AAH96085.1 | |||
BC099624.1 | AAH99624.1 | |||
BC099625.1 | AAH99625.1 | |||
BC111763.1 | AAI11764.1 | |||
BC112352.1 | AAI12353.1 | |||
BC113018.1 | ||||
BC113019.1 | ||||
BC128031.1 | ||||
M60315.1 | AAA36737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830582 | Chr.6:7706847 - 7889468 on Build GRCh38 | Loss | BMP6 BLOC1S5-TXNDC5 TXNDC5 |
esv3890787 | Chr.6:6996928 - 7981438 on Build GRCh38 | Loss | BMP6 BLOC1S5-TXNDC5 RREB1 SNRNP48 CAGE1 LOC101928076 TXNDC5 DSP RIOK1 SSR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |