Assay Details
Target Gene Details
Entrez Gene ID: | 11116 |
Gene Name: | FGFR1 oncogene partner |
Gene Aliases: |
FOP |
Location: |
Chr.6:166999317-167042418 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGFR1OP | NM_001278690.1 | NP_001265619.1 | ||
NM_007045.3 | NP_008976.1 | |||
NM_194429.2 | NP_919410.1 | |||
AJ420472.1 | ||||
AK289846.1 | ||||
AK294950.1 | ||||
AK312791.1 | ||||
BC011902.2 | AAH11902.1 | |||
DC418208.1 | ||||
Y18046.1 | CAA77020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv605260 | Chr.6:166570606 - 167041651 on Build GRCh38 | Gain | MIR3939 RNASET2 RPS6KA2-AS1 FGFR1OP RPS6KA2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |