Assay Details
Target Gene Details
Entrez Gene ID: | 222611 |
Gene Name: | adhesion G protein-coupled receptor F2 |
Gene Aliases: |
GPR111, PGR20, hGPCR35 |
Location: |
Chr.6:47656590-47697797 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRF2 | NM_153839.6 | NP_722581.4 | ||
AY140953.1 | AAN46667.1 | |||
DQ315369.1 | ABC41928.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26618 | Chr.6:47654534 - 47685135 on Build GRCh38 | Gain | ADGRF2 |
nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain | LOC101926962 TNFRSF21 ADGRF1 OPN5 ADGRF5 ADGRF2 PTCHD4 CD2AP ADGRF4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |