Assay Details
Target Gene Details
Entrez Gene ID: | 8464 |
Gene Name: | SPT3 homolog, SAGA and STAGA complex component |
Gene Aliases: |
SPT3, SPT3L |
Location: |
Chr.6:44808844-45378051 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SUPT3H |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5285 | Chr.6:44785336 - 44820455 on Build GRCh38 | Insertion | LOC101929770 SUPT3H |