Assay Details
Target Gene Details
Entrez Gene ID: | 55173 |
Gene Name: | mitochondrial ribosomal protein S10 |
Gene Aliases: |
MRP-S10, PNAS-122 |
Location: |
Chr.6:42206801-42217907 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPS10 | NM_018141.3 | NP_060611.2 | ||
XM_005249206.2 | XP_005249263.1 | |||
XM_011514724.2 | XP_011513026.1 | |||
XM_011514725.2 | XP_011513027.1 | |||
XM_017010996.1 | XP_016866485.1 | |||
AF113220.1 | AAG39291.1 | |||
AF277180.1 | AAK07539.1 | |||
AK001429.1 | BAA91686.1 | |||
AK098446.1 | ||||
AK298219.1 | ||||
AK316599.1 | ||||
BC012560.1 | AAH12560.1 | |||
BG776014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830649 | Chr.6:42047596 - 42231697 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)