Assay Details
Target Gene Details
Entrez Gene ID: | 643749 |
Gene Name: | TRAF3IP2 antisense RNA 1 |
Gene Aliases: |
C6UAS, C6orf3, NCRNA00248, TRAF3IP2-AS2 |
Location: |
Chr.6:111483472-111602295 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRAF3IP2-AS1 | NR_034108.1 | |||
NR_034109.1 | ||||
NR_034110.1 | ||||
NR_034111.1 | ||||
BX647114.1 |