Assay Details
Target Gene Details
Entrez Gene ID: | 55604 |
Gene Name: | capping protein regulator and myosin 1 linker 1 |
Gene Aliases: |
CARMIL, CARMIL1a, LRRC16, LRRC16A, dJ501N12.1, dJ501N12.5 |
Location: |
Chr.6:25279428-25620530 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CARMIL1 | NM_001173977.1 | NP_001167448.1 | ||
NM_017640.5 | NP_060110.4 | |||
XM_005249218.1 | XP_005249275.1 | |||
XM_017011008.1 | XP_016866497.1 | |||
XM_017011009.1 | XP_016866498.1 | |||
XM_017011010.1 | XP_016866499.1 | |||
XM_017011011.1 | XP_016866500.1 | |||
XM_017011012.1 | XP_016866501.1 | |||
XM_017011013.1 | XP_016866502.1 | |||
XM_017011014.1 | XP_016866503.1 | |||
XM_017011015.1 | XP_016866504.1 | |||
XM_017011016.1 | XP_016866505.1 | |||
XM_017011017.1 | XP_016866506.1 | |||
AK125696.1 | BAC86250.1 | |||
BC040903.1 | AAH40903.1 | |||
BC140770.1 | ||||
CD691491.1 | ||||
FJ009082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1025694 | Chr.6:25136632 - 25309222 on Build GRCh38 | Gain | CMAHP CARMIL1 LOC105375106 LOC101928663 |
esv3890801 | Chr.6:25274417 - 25428726 on Build GRCh38 | Gain | CARMIL1 LOC105375106 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |