Assay Details
Target Gene Details
Entrez Gene ID: | 23033 |
Gene Name: | dopey family member 1 |
Gene Aliases: |
DOP1, KIAA1117, dJ202D23.2 |
Location: |
Chr.6:83067650-83171351 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3609773 | Chr.6:83088352 - 83091427 on Build GRCh38 | Loss | DOPEY1 |
nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain | RWDD2A PGM3 DOPEY1 UBE3D |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |