Assay Details
Target Gene Details
Entrez Gene ID: | 26054 |
Gene Name: | SUMO1/sentrin specific peptidase 6 |
Gene Aliases: |
SSP1, SUSP1 |
Location: |
Chr.6:75601509-75718285 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SENP6 | NM_001100409.2 | NP_001093879.1 | ||
NM_001304792.1 | NP_001291721.1 | |||
NM_015571.3 | NP_056386.2 | |||
XM_006715457.3 | XP_006715520.1 | |||
XM_011535746.2 | XP_011534048.1 | |||
XM_011535747.2 | XP_011534049.1 | |||
XM_017010719.1 | XP_016866208.1 | |||
XM_017010722.1 | XP_016866211.1 | |||
XM_017010723.1 | XP_016866212.1 | |||
XM_017010724.1 | XP_016866213.1 | |||
XM_017010725.1 | XP_016866214.1 | |||
AB018340.2 | ||||
AF196304.1 | AAF04852.1 | |||
AF306508.1 | AAG30253.1 | |||
AF307849.1 | AAG29831.1 | |||
AK096455.1 | BAC04794.1 | |||
AK126921.1 | ||||
BC028583.1 | AAH28583.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)