Assay Details
Target Gene Details
Entrez Gene ID: | 135228 |
Gene Name: | CD109 molecule |
Gene Aliases: |
CPAMD7, p180, r150 |
Location: |
Chr.6:73679192-73828317 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD109 | XM_011535473.1 | XP_011533775.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576178 | Chr.6:73593218 - 73711991 on Build GRCh38 | Gain | SLC17A5 LOC101928489 CD109 |
dgv6015n100 | Chr.6:73590773 - 73714632 on Build GRCh38 | Gain | SLC17A5 LOC101928489 CD109 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |