Assay Details
Target Gene Details
Entrez Gene ID: | 221477 |
Gene Name: | chromosome 6 open reading frame 89 |
Gene Aliases: |
BRAP |
Location: |
Chr.6:36834886-36928964 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C6orf89 | XM_017010434.1 | XP_016865923.1 |
Target Gene Details
Entrez Gene ID: | 57699 |
Gene Name: | copine 5 |
Gene Aliases: |
COPN5, CPN5 |
Location: |
Chr.6:36740773-36839444 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CPNE5 | NM_001314017.1 | NP_001300946.1 | ||
NM_020939.1 | NP_065990.1 | |||
XM_005249247.2 | XP_005249304.1 | |||
XM_011514768.1 | XP_011513070.1 | |||
XM_011514769.1 | XP_011513071.1 | |||
XM_011514770.1 | XP_011513072.1 | |||
XM_011514772.1 | XP_011513074.1 | |||
AB046819.1 | ||||
DA265730.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5262 | Chr.6:36831553 - 36858434 on Build GRCh38 | Deletion | PPIL1 C6orf89 CPNE5 |
nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain | BTBD9 LOC107986531 SRSF3 PI16 C6orf89 GLP1R MDGA1 CCDC167 PPIL1 FGD2 LOC100505530 TMEM217 RAB44 CPNE5 PANDAR CDKN1A RNF8 MTCH1 ZFAND3 DNAH8 CMTR1 LOC100131047 PIM1 TBC1D22B GLO1 MIR3925 MIR4462 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |