Assay Details
Target Gene Details
Entrez Gene ID: | 51167 |
Gene Name: | cytochrome b5 reductase 4 |
Gene Aliases: |
NCB5OR, cb5/cb5R, dJ676J13.1 |
Location: |
Chr.6:83859651-83960427 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYB5R4 | NM_016230.3 | NP_057314.2 | ||
AF169803.1 | AAF04812.1 | |||
AK313146.1 | ||||
AL832113.1 | ||||
BC025380.1 | AAH25380.2 | |||
DA274558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830718 | Chr.6:83781810 - 83941347 on Build GRCh38 | Gain+Loss | CYB5R4 RIPPLY2 |
nsv1025798 | Chr.6:83654421 - 84334553 on Build GRCh38 | Gain | CEP162 MRAP2 SNAP91 LOC105377879 CYB5R4 RIPPLY2 |
esv2669183 | Chr.6:83873000 - 83879169 on Build GRCh38 | Deletion | CYB5R4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |