Assay Details
Target Gene Details
Entrez Gene ID: | 221442 |
Gene Name: | adenylate cyclase 10, soluble pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.6:41101034-41140835 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADCY10P1 | NR_026938.2 | |||
BX537595.1 | ||||
BX648193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv602970 | Chr.6:41070039 - 41258521 on Build GRCh38 | Gain | TREML1 ADCY10P1 TREML4 NFYA TREML2 LOC105375056 TREML5P TREM2 OARD1 TREML3P |