Assay Details
Target Gene Details
Entrez Gene ID: | 114801 |
Gene Name: | transmembrane protein 200A |
Gene Aliases: |
KIAA1913, TTMA, TTMC |
Location: |
Chr.6:130366281-130443424 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM200A | NM_001258276.1 | NP_001245205.1 | ||
NM_001258277.1 | NP_001245206.1 | |||
XM_005266816.2 | XP_005266873.1 | |||
XM_011535419.2 | XP_011533721.1 | |||
XM_017010230.1 | XP_016865719.1 | |||
AB067500.1 | ||||
AK315824.1 | ||||
BC044246.1 | AAH44246.1 | |||
DA657363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv604663 | Chr.6:129482340 - 130671010 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)