Assay Details
Target Gene Details
Entrez Gene ID: | 50508 |
Gene Name: | NADPH oxidase 3 |
Gene Aliases: |
GP91-3, MOX-2 |
Location: |
Chr.6:155395368-155455903 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NOX3 | NM_015718.2 | NP_056533.1 | ||
AF190122.1 | AAG17121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1025003 | Chr.6:155077125 - 155755140 on Build GRCh38 | Gain | CLDN20 TIAM2 NOX3 LOC105378068 TFB1M |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |